CAS RN 58-61-7
Thermo Scientific Chemicals Adenosine, 99%
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 IUPAC Name: (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Thermo Scientific Chemicals Adenosine, 99+%
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 IUPAC Name: (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine, 99.8%, MP Biomedicals™
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine, MP Biomedicals
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
phyproof™ Adenosine, ≥98% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Adenosine, MP Biomedicals™
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Adenosine 99.0+%, TCI America™
CAS: 58-61-7 Molecular Formula: C10H13N5O4 Molecular Weight (g/mol): 267.25 MDL Number: MFCD00005752 InChI Key: OIRDTQYFTABQOQ-KQYNXXCUSA-N Synonym: adenosine,adenocard,adenoscan,adenine riboside,adenosin,beta-d-adenosine,nucleocardyl,boniton,sandesin,myocol PubChem CID: 60961 ChEBI: CHEBI:16335 IUPAC Name: (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1