Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine, MP Biomedicals

Catalog No. MP219980380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219980380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219980380 Supplier MP Biomedicals, Inc Supplier No. 0219980380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Adenosine is used in topical treatment of foot wounds in diabetes mellitus and also used for the treatment or prevention of cardiac arrhythmias (irregular heartbeat)
  • It is used to cause dilation of the blood vessels

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Melting Point 233°C to 238°C
Color White
Linear Formula C10H13N5O4
Quantity 100 g
Formula Weight 267.2
Grade USP
Physical Form Powder
Chemical Name or Material ADENOSINE
Recommended Storage -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.