accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH2NNNH2S

CAS RN 581-64-6

IUPAC Name: 3,7-diamino-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: cyanine lauthsches violett thionine lauth's violet thionin 硫堇 katalysin
Molecular Weight (g/mol): 263.74
Molecular Formula: C12H10ClN3S
InChi Key: ANRHNWWPFJCPAZ-UHFFFAOYSA-M
SMILES: [Cl-].NC1=CC2=[S+]C3=CC(N)=CC=C3N=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)