CAS RN 582315-72-8
IUPAC Name:
4-butoxy-5-(2,6-difluoro-4-methylbenzoyl)-1H-pyrazolo[3,4-b]pyridine
Synonyms:
4-butoxy-5-(2,6-difluoro-4-methylbenzoyl)-1H-pyrazolo[3,4-b]pyridine
Molecular Weight (g/mol):
345.35
Molecular Formula:
C18H17F2N3O2
InChi Key:
SCFMWQIQBVZOQR-UHFFFAOYSA-N
SMILES:
CCCCOC1=C2C=NNC2=NC=C1C(=O)C1=C(F)C=C(C)C=C1F
1
–
1
of
1
results
1
Tocris Bioscience™ BMS 265246, >97%
CAS: 582315-72-8 Molecular Formula: C18H17F2N3O2