accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSSNO

CAS RN 587871-26-9

IUPAC Name: 2-(morpholin-4-yl)-6-(thianthren-1-yl)-4H-pyran-4-one
Synonyms: 2-(morpholin-4-yl)-6-(thianthren-1-yl)pyran-4-one
Molecular Weight (g/mol): 395.49
Molecular Formula: C21H17NO3S2
InChi Key: XRKYMMUGXMWDAO-UHFFFAOYSA-N
SMILES: O=C1C=C(OC(=C1)C1=CC=CC2=C1SC1=CC=CC=C1S2)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)