CAS RN 59-92-7
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%
CAS: 59-92-7 Molecular Formula: C9H11NO4 Molecular Weight (g/mol): 197.19 MDL Number: MFCD00002598 InChI Key: WTDRDQBEARUVNC-LURJTMIESA-N Synonym: levodopa,l-dopa,dopar,3-hydroxy-l-tyrosine,3,4-dihydroxy-l-phenylalanine,larodopa,bendopa,levopa,dihydroxy-l-phenylalanine,cidandopa PubChem CID: 6047 ChEBI: CHEBI:15765 IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
3-(3,4-Dihydroxyphenyl)-L-alanine 98.0+%, TCI America™
CAS: 59-92-7 Molecular Formula: C9H11NO4 Molecular Weight (g/mol): 197.19 MDL Number: MFCD00002598 InChI Key: WTDRDQBEARUVNC-LURJTMIESA-N Synonym: levodopa,l-dopa,dopar,3-hydroxy-l-tyrosine,3,4-dihydroxy-l-phenylalanine,larodopa,bendopa,levopa,dihydroxy-l-phenylalanine,cidandopa PubChem CID: 6047 ChEBI: CHEBI:15765 IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
L-β-3,4-Dihydroxyphenylalanine, 99%, MP Biomedicals™
CAS: 59-92-7 Molecular Formula: C9H11NO4 Molecular Weight (g/mol): 197.19 MDL Number: MFCD00002598 InChI Key: WTDRDQBEARUVNC-LURJTMIESA-N Synonym: levodopa,l-dopa,dopar,3-hydroxy-l-tyrosine,3,4-dihydroxy-l-phenylalanine,larodopa,bendopa,levopa,dihydroxy-l-phenylalanine,cidandopa PubChem CID: 6047 ChEBI: CHEBI:15765 IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
Levodopa, Powder, USP, 98-102%, Spectrum™ Chemical
CAS: 59-92-7 Molecular Formula: C9H11NO4 Molecular Weight (g/mol): 197.19 MDL Number: MFCD00002598 InChI Key: WTDRDQBEARUVNC-LURJTMIESA-N IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
Thermo Scientific Chemicals 3,4-Dihydroxy-L-phenylalanine, 98+%
CAS: 59-92-7 Molecular Formula: C9H11NO4 Molecular Weight (g/mol): 197.19 MDL Number: MFCD00002598 InChI Key: WTDRDQBEARUVNC-LURJTMIESA-N Synonym: levodopa,l-dopa,dopar,3-hydroxy-l-tyrosine,3,4-dihydroxy-l-phenylalanine,larodopa,bendopa,levopa,dihydroxy-l-phenylalanine,cidandopa PubChem CID: 6047 ChEBI: CHEBI:15765 IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
R&D Systems™ L-DOPA
CAS: 59-92-7 Molecular Formula: C9H11NO4