accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59099-58-0

CH3OOSOOFFF

CAS RN 59099-58-0

IUPAC Name: 2-methoxyphenyl trifluoromethanesulfonate
Synonyms: 2-methoxyphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 256.2
Molecular Formula: C8H7F3O4S
InChi Key: RCDZTJFBROIDKD-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results