accessibility menu, dialog, popup

UserName

Viewing profile as user:
Pb

CAS RN 595-89-1

IUPAC Name: tetraphenylplumbane
Synonyms: tetraphenyllead PbPh4 tetraphenyl lead
Molecular Weight (g/mol): 515.6
Molecular Formula: C24H20Pb
InChi Key: WBJSMHDYLOJVKC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)