Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenyllead 97%, Thermo Scientific™

Catalog No. AC138560100
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC138560100 10 g
AC138560050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138560100 Supplier Thermo Scientific Chemicals Supplier No. 138560100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-89-1
Molecular Formula C24H20Pb
Molecular Weight (g/mol) 515.60
MDL Number MFCD00002999
InChI Key WBJSMHDYLOJVKC-UHFFFAOYSA-N
Synonym tetraphenyllead, tetraphenyl lead, plumbane, tetraphenyl, pbph4, lead, tetraphenyl-,, lead tetraphenyl, acmc-209mel, wbjsmhdylojvkc-uhfffaoysa
PubChem CID 72906
ChEBI CHEBI:30184
IUPAC Name tetraphenylplumbane
SMILES C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 228°C to 231°C
Color White
Infrared Spectrum Authentic
Linear Formula Pb(C6H5)4
Quantity 10 g
Beilstein 16, 917
Formula Weight 515.6
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Tetraphenyllead, 97%
EINECSNumber 209-871-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.