Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenyllead, 97%, Thermo Scientific™

Catalog No. AAA1220822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1220822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1220822 Supplier Thermo Scientific Chemicals Supplier No. A1220822

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-89-1
Molecular Formula C24H20Pb
Molecular Weight (g/mol) 515.60
MDL Number MFCD00002999
InChI Key WBJSMHDYLOJVKC-UHFFFAOYSA-N
Synonym tetraphenyllead, tetraphenyl lead, plumbane, tetraphenyl, pbph4, lead, tetraphenyl-,, lead tetraphenyl, acmc-209mel, wbjsmhdylojvkc-uhfffaoysa
PubChem CID 72906
ChEBI CHEBI:30184
IUPAC Name tetraphenylplumbane
SMILES C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C to 228°C
Boiling Point 126°C (13 mmHg)
Quantity 100 g
UN Number UN3467
Formula Weight 515.61
Percent Purity 97%
Chemical Name or Material Tetraphenyllead
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
EINECSNumber 209-871-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.