CAS RN 59865-13-3
Thermo Scientific Chemicals Cyclosporin A, 98%
CAS: 59865-13-3 Molecular Formula: C62H111N11O12 Molecular Weight (g/mol): 1202.64 MDL Number: MFCD00274558 InChI Key: PMATZTZNYRCHOR-IMVLJIQENA-N PubChem CID: 132274082 SMILES: CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
Cyclosporin A, 99+%
CAS: 59865-13-3 Molecular Formula: C62H111N11O12 Molecular Weight (g/mol): 1202.64 MDL Number: MFCD00274558 InChI Key: PMATZTZNYRCHOR-IMVLJIQENA-N Synonym: Cyclosporine A; Antibiotic S 7481F1 PubChem CID: 132274082 IUPAC Name: 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone SMILES: CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
Cyclosporine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Cyclosporin A 97.0+%, TCI America™
CAS: 59865-13-3 Molecular Formula: C62H111N11O12 Molecular Weight (g/mol): 1202.64 MDL Number: MFCD00274558 InChI Key: PMATZTZNYRCHOR-IMVLJIQENA-N Synonym: cyclosporin a PubChem CID: 132274082 IUPAC Name: 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone SMILES: CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
R&D Systems™ Cyclosporin A
CAS: 59865-13-3 Molecular Formula: C62H111N11O12
Medisca Cyclosporine A USP
Cyclosporine A USP is a white to almost white powder. Alternative name(s): ciclosporin, cyclosporin, Cyclosporin A