accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CH3CNNNHSOOCH3

CAS RN 602306-29-6

IUPAC Name: N-(4-methanesulfonylphenyl)-4-[2-methyl-1-(propan-2-yl)-1H-imidazol-5-yl]pyrimidin-2-amine
Synonyms: 4-(3-isopropyl-2-methylimidazol-4-yl)-N-(4-methanesulfonylphenyl)pyrimidin-2-amine
Molecular Weight (g/mol): 371.46
Molecular Formula: C18H21N5O2S
InChi Key: WJRRGYBTGDJBFX-UHFFFAOYSA-N
SMILES: CC(C)N1C(C)=NC=C1C1=NC(NC2=CC=C(C=C2)S(C)(=O)=O)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results