CAS RN 603151-83-3
IUPAC Name:
9-(cyclopropylmethyl)-3-(2,4-dichlorophenyl)-10-ethyl-6-methyl-1,2,5,9-tetraazatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene hydrochloride
Synonyms:
9-(cyclopropylmethyl)-3-(2,4-dichlorophenyl)-10-ethyl-6-methyl-1,2,5,9-tetraazatricyclo[6.3.1.0⁴,¹²]dodeca-2,4,6,8(12)-tetraene hydrochloride
Molecular Weight (g/mol):
437.79
Molecular Formula:
C21H23Cl3N4
InChi Key:
LNYUWBPFYXPUIO-UHFFFAOYNA-N
SMILES:
Cl.CCC1CN2N=C(C3=NC(C)=CC(N1CC1CC1)=C23)C1=C(Cl)C=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ NBI 35965 hydrochloride
CAS: 603151-83-3 Molecular Formula: C21H23Cl3N4