Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NBI 35965 hydrochloride
GREENER_CHOICESDP

Catalog No. 310050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-005-0 50 mg
31-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-005-0 Supplier R&D Systems™ Supplier No. 3100/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective CRF1 antagonist

Potent and selective corticotropin-releasing factor receptor 1 (CRF1) antagonist (Ki values are 4 and > 10000 nM at CRF1 and CRF2 respectively). Inhibits cAMP accumulation and adrenocorticotropic hormone (ACTH) production in vitro (pIC50 values are 7.1 and 6.9 respectively) and attenuates CRF and ACTH production in vivo. Orally active and brain penetrant with anxiolytic activity.

Chemical Identifiers

CAS 603151-83-3
Molecular Formula C21H23Cl3N4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 437.79
Purity >98%
Molecular Weight (g/mol) 437.793
Chemical Name or Material NBI 35965 hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.