accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6044-68-4

IUPAC Name: 3,3-dimethoxyprop-1-ene
Synonyms: 3,3-dimethoxy-propene
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: OBWGMYALGNDUNM-UHFFFAOYSA-N
SMILES: COC(OC)C=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)