Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein Dimethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. A245525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A245525G 25 g
A2455250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A245525G Supplier TCI America Supplier No. A245525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6044-68-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008632
InChI Key OBWGMYALGNDUNM-UHFFFAOYSA-N
Synonym 3,3-Dimethoxy-1-propene
PubChem CID 80145
IUPAC Name 3,3-dimethoxyprop-1-ene
SMILES COC(C=C)OC

Specifications

Color Colorless
Boiling Point 90°C
Quantity 25 g
UN Number 1993
Formula Weight 102.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acrolein Dimethyl Acetal
RTECSNumber UC8500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.