CAS RN 607378-18-7
IUPAC Name:
2-(adamantan-1-yl)-N-[2-({2-[(2-hydroxyethyl)amino]ethyl}amino)quinolin-5-yl]acetamide dihydrochloride
Synonyms:
2-(adamantan-1-yl)-N-[2-({2-[(2-hydroxyethyl)amino]ethyl}amino)quinolin-5-yl]acetamide dihydrochloride
Molecular Weight (g/mol):
495.49
Molecular Formula:
C25H36Cl2N4O2
InChi Key:
BVFONFUUWORSPO-UHFFFAOYSA-N
SMILES:
Cl.Cl.OCCNCCNC1=NC2=CC=CC(NC(=O)CC34CC5CC(CC(C5)C3)C4)=C2C=C1
1
–
1
of
1
results
1
R&D Systems™ AZ 10606120 dihydrochloride
CAS: 607378-18-7 Molecular Formula: C25H36Cl2N4O2