accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60835-71-4

IUPAC Name: 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin-15-amine
Synonyms: 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin-15-amine
Molecular Weight (g/mol): 283.32
Molecular Formula: C14H21NO5
InChi Key: CQNGAZMLFIMLQN-UHFFFAOYSA-N
SMILES: NC1=CC2=C(OCCOCCOCCOCCO2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)