Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Aminobenzo-15-crown 5-Ether 97.0+%, TCI America™
SDP

Catalog No. A14041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14041G Supplier TCI America Supplier No. A14041G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60835-71-4
Molecular Formula C14H21NO5
Molecular Weight (g/mol) 283.324
MDL Number MFCD00068686
InChI Key CQNGAZMLFIMLQN-UHFFFAOYSA-N
Synonym 2,3-(4-Aminobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 2724797
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-trien-17-amine
SMILES C1COCCOC2=C(C=C(C=C2)N)OCCOCCO1

Specifications

Melting Point 79°C
Quantity 1 g
Formula Weight 283.32
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminobenzo-15-crown 5-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.