accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOPOOONNH2NNNOONOONOONOOOPHOOOPOOO

CAS RN 61368-63-6

IUPAC Name: 9-[4'-({[hydroxy({[hydroxy(phosphonatooxy)phosphoryl]oxy})phosphoryl]oxy}methyl)-2,4,6-trinitrospiro[cyclohexane-1,2'-furo[3,4-d][1,3]dioxole]-2,4-dien-6'-yl]-6,9-dihydro-5H-purin-6-iminium
Synonyms: 9-[4'-({[hydroxy([hydroxy(phosphonatooxy)phosphoryl]oxy)phosphoryl]oxy}methyl)-2,4,6-trinitrospiro[cyclohexane-1,2'-furo[3,4-d][1,3]dioxole]-2,4-dien-6'-yl]-5H-purin-6-iminium
Molecular Weight (g/mol): 713.23
Molecular Formula: C16H12N8O19P3
InChi Key: SWLXZTUOMCLGJI-UHFFFAOYNA-M
SMILES: OP(=O)(OCC1=C2OC3(OC2=C(O1)N1C=NC2C1=NC=NC2=[NH2+])C(C=C(C=C3[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)OP(O)(=O)OP([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results