Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TNP-ATP triethylammonium salt
GREENER_CHOICESDP

Catalog No. 24645
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-645 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-645 Supplier Tocris Bioscience™ Supplier No. 2464/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective P2X antagonist

High affinity, selective P2X receptor antagonist. Inhibits ATP-induced currents in cells expressing P2X1, P2X3 and heteromeric P2X2/3 receptors with IC50 values of 6, 0.9 and 7 nM respectively. Displays 1000-fold selectivity over P2X2, P2X4 and P2X7.

Chemical Identifiers

CAS 61368-63-6
Molecular Formula C16H13N8O19P3·C24H64N

Specifications

Quantity 5 mg
Formula Weight 1123.04
Purity >95%
Chemical Name or Material TNP-ATP triethylammonium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.