Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylaminesulfonic Acid Sodium Salt (Crystalline/Certified ACS), Fisher Chemical

Catalog No. D1415 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D141-5 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D141-5

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) 272.27
MDL Number MFCD00036468
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
PubChem CID 517002
IUPAC Name 4-(phenylamino)benzene-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 200°C
Color Yellow
Packaging Glass Bottle
Quantity 5 g
Sensitivity Pass Test
Formula Weight 271.27
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Diphenylaminesulfonic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.