accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61699-62-5

IUPAC Name: bis(propan-2-yloxy)cyclobut-3-ene-1,2-dione
Synonyms: diisopropoxycyclobut-3-ene-1,2-dione
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: KCZPGGVPQXQGEJ-UHFFFAOYSA-N
SMILES: CC(C)OC1=C(OC(C)C)C(=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results