accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62-73-7

IUPAC Name: 2,2-dichloroethenyl dimethyl phosphate
Synonyms: dichlorvos herkal marvex chlorvinphos nuvan 100EC herkol mopari deriban vapona insecticide lindanmafu
Molecular Weight (g/mol): 220.97
Molecular Formula: C4H7Cl2O4P
InChi Key: OEBRKCOSUFCWJD-UHFFFAOYSA-N
SMILES: COP(=O)(OC)OC=C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results