accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3N

CAS RN 6231-18-1

IUPAC Name: 2,6-dimethoxypyridine
Synonyms: pyridine, 2,6-dimethoxy-
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: IBTGEEMBZJBBSH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=N1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results