accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623142-96-1

IUPAC Name: 1-{4-[(6,7-dimethoxyquinolin-4-yl)oxy]-2-methoxyphenyl}-3-[1-(1,3-thiazol-2-yl)ethyl]urea
Synonyms: 1-{4-[(6,7-dimethoxyquinolin-4-yl)oxy]-2-methoxyphenyl}-3-[1-(1,3-thiazol-2-yl)ethyl]urea
Molecular Weight (g/mol): 480.54
Molecular Formula: C24H24N4O5S
InChi Key: SHPFDGWALWEPGS-UHFFFAOYNA-N
SMILES: COC1=C(NC(=O)NC(C)C2=NC=CS2)C=CC(OC2=CC=NC3=CC(OC)=C(OC)C=C23)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)