CAS RN 627536-09-8
IUPAC Name:
2-(5-chloro-2-fluorophenyl)-N-(pyridin-4-yl)pteridin-4-amine
Synonyms:
2-(5-chloro-2-fluorophenyl)-N-(pyridin-4-yl)pteridin-4-amine
Molecular Weight (g/mol):
352.76
Molecular Formula:
C17H10ClFN6
InChi Key:
BERLXWPRSBJFHO-UHFFFAOYSA-N
SMILES:
FC1=C(C=C(Cl)C=C1)C1=NC(NC2=CC=NC=C2)=C2N=CC=NC2=N1
1
–
2
of
2
results
1
R&D Systems™ SD 208
CAS: 627536-09-8 Molecular Formula: C17H10ClFN6
2
MilliporeSigma™ Calbiochem™ TGF-β RI Kinase Inhibitor V
The TGF-β RI Kinase Inhibitor V, also referenced under CAS 627536-09-8, controls the biological activity of TGF-β RI. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.