accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63071-03-4

IUPAC Name: 2-methoxy-6-methylpyridine
Synonyms: 2-methoxy-6-methylpyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: WIMNZMOEBDPZTB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)