Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-6-methylpyridine, 98%

Catalog No. AAH2577206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2577206 5 g
AAH2577203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2577206 Supplier Thermo Scientific Chemicals Supplier No. H2577206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 63071-03-4 | C7H9NO | 123.155 g/mol

2-Methoxy-6-methylpyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxy-6-methylpyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63071-03-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD04112507
InChI Key WIMNZMOEBDPZTB-UHFFFAOYSA-N
Synonym 2-methoxy-6-methyl-pyridine, 2-methoxy-6-picoline, 6-methoxy-2-picoline, 2-methoxy-6-methylam, acmc-209yyt, 6-methoxy-2-methylpyridine, 2-methoxy-6-methyl pyridine, ksc490s3b, 2-methyl-6-methyloxy pyridine, 2-methoxy-6-methylpyridine 1g
PubChem CID 5324773
IUPAC Name 2-methoxy-6-methylpyridine
SMILES CC1=NC(=CC=C1)OC

Specifications

Density 1.01
Boiling Point 156°C
Flash Point 49°C (120°F)
Refractive Index 1.4995
Quantity 5 g
UN Number UN1993
Solubility Information Slightly soluble in water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Methoxy-6-methylpyridine
Hazard Category H226-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.