Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-6-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. M16955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16955G 5 g
M169525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16955G Supplier TCI America Supplier No. M16955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63071-03-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
InChI Key WIMNZMOEBDPZTB-UHFFFAOYSA-N
Synonym 2-methoxy-6-methyl-pyridine, 2-methoxy-6-picoline, 6-methoxy-2-picoline, 2-methoxy-6-methylam, acmc-209yyt, 6-methoxy-2-methylpyridine, 2-methoxy-6-methyl pyridine, ksc490s3b, 2-methyl-6-methyloxy pyridine, 2-methoxy-6-methylpyridine 1g
PubChem CID 5324773
IUPAC Name 2-methoxy-6-methylpyridine
SMILES CC1=NC(=CC=C1)OC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 g
UN Number 1993
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-6-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.