accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 634908-75-1

IUPAC Name: N-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-N-(pyridin-2-yl)cyclohexanecarboxamide; formic acid; prop-2-enoic acid
Synonyms: N-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-N-(pyridin-2-yl)cyclohexanecarboxamide; acrylic acid; formic acid
Molecular Weight (g/mol): 540.66
Molecular Formula: C29H40N4O6
InChi Key: QYOBEWLYELTWTQ-UHFFFAOYSA-N
SMILES: OC=O.OC(=O)C=C.COC1=CC=CC=C1N1CCN(CCN(C(=O)C2CCCCC2)C2=CC=CC=N2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results