CAS RN 636-00-0
IUPAC Name:
hydrogen 5-(2-aminoethyl)benzene-1,2,4-triol bromide
Synonyms:
hydrogen hydroxydopamine bromide
Molecular Weight (g/mol):
250.09
Molecular Formula:
C8H12BrNO3
InChi Key:
MLACDGUOKDOLGC-UHFFFAOYSA-N
SMILES:
[H+].[Br-].NCCC1=CC(O)=C(O)C=C1O
1
–
1
of
1
results
1
R&D Systems™ 6-Hydroxydopamine hydrobromide
CAS: 636-00-0 Molecular Formula: C8H12BrNO3