accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNNH2(E/Z)

CAS RN 6416-57-5

IUPAC Name: 4-[2-(naphthalen-1-yl)diazen-1-yl]benzene-1,3-diamine
Synonyms: 4-[2-(naphthalen-1-yl)diazen-1-yl]benzene-1,3-diamine
Molecular Weight (g/mol): 262.32
Molecular Formula: C16H14N4
InChi Key: AHWMWMNEYBHQNL-UHFFFAOYSA-N
SMILES: NC1=CC=C(N=NC2=C3C=CC=CC3=CC=C2)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results