accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOHOHOHHO

CAS RN 64395-92-2

IUPAC Name: 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl dodecanoate
Synonyms: 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl dodecanoate
Molecular Weight (g/mol): 362.46
Molecular Formula: C18H34O7
InChi Key: HABWUWJGNVZVPU-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(=O)OC1OC(CO)C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results