accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6508-43-6

IUPAC Name: (9S)-9-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3,3-dimethyl-2,3,4,9-tetrahydro-1H-xanthen-1-one
Synonyms: (9S)-9-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3,3-dimethyl-4,9-dihydro-2H-xanthen-1-one
Molecular Weight (g/mol): 366.46
Molecular Formula: C23H26O4
InChi Key: CCJIUBMPDVWYLU-IBGZPJMESA-N
SMILES: CC1(C)CC(O)=C([C@@H]2C3=CC=CC=C3OC3=C2C(=O)CC(C)(C)C3)C(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results