accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65192-28-1

H3COOHO(Z)

CAS RN 65192-28-1

IUPAC Name: (2Z)-3-hydroxy-2-(4-methoxyphenyl)prop-2-enal
Synonyms: (2Z)-3-hydroxy-2-(4-methoxyphenyl)prop-2-enal
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: WASQXFIYEDZPDK-RMKNXTFCSA-N
SMILES: COC1=CC=C(C=C1)C(=C\O)\C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results