CAS RN 652973-93-8
IUPAC Name:
4-chloro-N-(2-{[1-(naphthalen-1-yl)ethyl]amino}cyclohexyl)benzamide
Synonyms:
4-chloro-N-(2-{[1-(naphthalen-1-yl)ethyl]amino}cyclohexyl)benzamide
Molecular Weight (g/mol):
406.95
Molecular Formula:
C25H27ClN2O
InChi Key:
YTFUQWWKTIWYEY-UHFFFAOYNA-N
SMILES:
CC(NC1CCCCC1NC(=O)C1=CC=C(Cl)C=C1)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
R&D Systems™ Calhex 231 hydrochloride
CAS: 652973-93-8 Molecular Formula: C25H27ClN2O