accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 654054-57-6

IUPAC Name: 2,9-dimethyl-1,10-phenanthroline hydrate
Synonyms: neocuproine hydrate
Molecular Weight (g/mol): 226.28
Molecular Formula: C14H14N2O
InChi Key: MFZBSWSCIWCRKS-UHFFFAOYSA-N
SMILES: O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)