accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONOONHOCH3

CAS RN 660846-41-3

IUPAC Name: tert-butyl 4-({[(2-methoxyphenyl)carbamoyl]oxy}methyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-({[(2-methoxyphenyl)carbamoyl]oxy}methyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 364.44
Molecular Formula: C19H28N2O5
InChi Key: SAXGSDIZIYFNKD-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1NC(=O)OCC1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results