accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOHOOHOHOH2N

CAS RN 66515-29-5

IUPAC Name: 2-amino-2-methyl-3-(phosphonooxy)propanoic acid
Synonyms: 2-amino-2-methyl-3-(phosphonooxy)propanoic acid
Molecular Weight (g/mol): 199.1
Molecular Formula: C4H10NO6P
InChi Key: GSFCOAGADOGIGE-UHFFFAOYNA-N
SMILES: CC(N)(COP(O)(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)