Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
MSOP is a selective group III metabotropic glutamate receptor antagonist. Displays an apparent KD of 51 μM for the L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord compared to > 700 μM for the (1S,3S)-ACPD-sensitive presynaptic mGlu in the same system. Has no activity on postsynaptic mGlu receptors or on ionotropic glutamate receptors on neonatal rat motoneurons.
Specifications
Specifications
| Chemical Name or Material | (RS)-α-Methylserine-O-phosphate |
| CAS | 66515-29-5 |
| Quantity | 5 mg |
| Target | Group III mGluR Antagonists |
| Molecular Formula | C4H10NO6P |
| Formula Weight | Observed MW: 199.1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

