CAS RN 67-03-8
Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5%
CAS: 67-03-8 Molecular Formula: C12H17ClN4OS·HCl Molecular Weight (g/mol): 337.26 MDL Number: MFCD00012780 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 MDL Number: MFCD00012780 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine Hydrochloride, Powder, MP Biomedicals
CAS: 67-03-8 Molecular Formula: C12H17ClN4OS·HCl Molecular Weight (g/mol): 337.263 MDL Number: MFCD00012780 Synonym: Aneurine hydrochloride,Vitamin B1 hydrochloride,Vitamin B1 HCl,vitamin b1 mononitrate,aneurine mononitrate,aneurine nitrate,betabion mononitrate,thiamine nitrate salt,vitamin b sub 1 nitrate,thiamine mononitrate usp
Thiamine Hydrochloride Hydrate 98.0+%, TCI America™
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 MDL Number: MFCD00012780 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards
Thiamine Hydrochloride, FCC, 98-102%, Spectrum™ Chemical
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.26 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M IUPAC Name: hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride SMILES: [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N
Thiamine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.26 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M IUPAC Name: hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride SMILES: [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N
Thiamine, Hydrochloride, MP Biomedicals
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine Hydrochloride (Crystalline), MP Biomedicals
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.26 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M IUPAC Name: hydrogen 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium dichloride SMILES: [H+].[Cl-].[Cl-].CC1=C(CCO)SC=[N+]1CC1=CN=C(C)N=C1N
Thiamine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.263 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,vitamin b1 hydrochloride,thiamine hcl,aneurine hydrochloride,berin,thiamine chloride hydrochloride,trophite,thiamin chloride,thiamine dichloride,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105 IUPAC Name: 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride SMILES: CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
Thiamine Hydrochloride, 100%, Cell Culture Reagent, MP Biomedicals™
CAS: 67-03-8 Molecular Formula: C12H18Cl2N4OS Molecular Weight (g/mol): 337.26 InChI Key: DPJRMOMPQZCRJU-UHFFFAOYSA-M Synonym: thiamine hydrochloride,aneurine hydrochloride,thiamine hcl,vitamin b1 hydrochloride,thiamine dichloride,thiamine chloride hydrochloride,trophite,thiamin chloride,vitamin b1,thiaminium chloride PubChem CID: 6202 ChEBI: CHEBI:49105