Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Hydrochloride, 100%, Cell Culture Reagent, MP Biomedicals™

Catalog No. ICN19474980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19474980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19474980 Supplier MP Biomedicals, Inc Supplier No. 0219474980
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.26
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, aneurine hydrochloride, thiamine hcl, vitamin b1 hydrochloride, thiamine dichloride, thiamine chloride hydrochloride, trophite, thiamin chloride, vitamin b1, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105

Specifications

Quantity 100 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.