accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)OHNOOOO

CAS RN 6729-55-1

IUPAC Name: (2R)-2-azaniumyl-4-[(carboxylatomethyl)carbamoyl]butanoate
Synonyms: (2R)-2-ammonio-4-(carboxylatomethylcarbamoyl)butanoate
Molecular Weight (g/mol): 203.18
Molecular Formula: C7H11N2O5
InChi Key: ACIJGUBIMXQCMF-SCSAIBSYSA-M
SMILES: [NH3+][C@H](CCC(=O)NCC([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)