CAS RN 675103-36-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BAY 678
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor