CAS RN 677338-12-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ PIK 90
Potent PI3K inhibitor, also used in hPSCs mesendoderm differentiation
2
MilliporeSigma™ Calbiochem™ PI 3-K inhibitor IX, PIK-90
The PI 3-K inhibitor IX, PIK-90, also referenced under CAS 677338-12-4, controls the biological activity of PI 3-K. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.