accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNSOOFFFFFSOOCl

CAS RN 677772-84-8

IUPAC Name: N-[4-(4-chlorobenzenesulfonyl)-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide
Synonyms: N-[4-(4-chlorobenzenesulfonyl)-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide
Molecular Weight (g/mol): 517.91
Molecular Formula: C19H17ClF5NO4S2
InChi Key: WDZVWDXOIGQJIO-UHFFFAOYSA-N
SMILES: FC1=CC(=C(F)C=C1)C1(CCC(CC1)NS(=O)(=O)C(F)(F)F)S(=O)(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)