CAS RN 68099-86-5
CAS RN 68099-86-5
IUPAC Name:
hydrogen N-benzyl-N-[3-(2-methylpropoxy)-2-(pyrrolidin-1-yl)propyl]aniline chloride
Synonyms:
bepridil hydrochloride
Molecular Weight (g/mol):
403.01
Molecular Formula:
C24H35ClN2O
InChi Key:
JXBBWYGMTNAYNM-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C)COCC(CN(CC1=CC=CC=C1)C1=CC=CC=C1)N1CCCC1
1
–
1
of
1
results
1
Tocris Bioscience™ Bepridil hydrochloride
CAS: 68099-86-5 Molecular Formula: C24H35ClN2O