accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3H3COHSOOO

CAS RN 68214-07-3

IUPAC Name: hexadecyltrimethylazanium hydrogen sulfate
Synonyms: cetyl-trimethyl-ammonium hydrogen sulfate
Molecular Weight (g/mol): 381.62
Molecular Formula: C19H43NO4S
InChi Key: UCRJJNVFJGKYQT-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)