CAS RN 6872-06-6
CAS RN 6872-06-6
IUPAC Name:
2-methyl-2,3-dihydro-1H-indole
Synonyms:
2-methyl-2,3-dihydro-1H-indole
Molecular Weight (g/mol):
133.19
Molecular Formula:
C9H11N
InChi Key:
QRWRJDVVXAXGBT-UHFFFAOYNA-N
SMILES:
CC1CC2=CC=CC=C2N1
1
–
2
of
2
results
1
Thermo Scientific Chemicals 2-Methylindoline, 99+%
CAS: 6872-06-6 Molecular Formula: C9H11N Molecular Weight (g/mol): 133.19 MDL Number: MFCD00005706 InChI Key: QRWRJDVVXAXGBT-UHFFFAOYNA-N Synonym: 2-methylindoline,2-methyl-2,3-dihydro-1h-indole,2,3-dihydro-2-methylindole,1h-indole, 2,3-dihydro-2-methyl,indoline, 2-methyl,2-methyl-2,3-dihydroindole,dihydroindole, 2-methyl,alpha-methyldihydroindole,.alpha.-methyldihydroindole,2,3-dihydro-2-methyl-1h-indole PubChem CID: 23305
2
2-Methylindoline 98.0+%, TCI America™
CAS: 6872-06-6 Molecular Formula: C9H11N Molecular Weight (g/mol): 133.19 MDL Number: MFCD00005706 InChI Key: QRWRJDVVXAXGBT-UHFFFAOYNA-N PubChem CID: 23305 IUPAC Name: 2-methyl-2,3-dihydro-1H-indole SMILES: CC1CC2=CC=CC=C2N1